3-Nitrosalicylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-3-nitrobenzoic acid3-Nitrosalicylic Acid

39982-0010 39982-0050
TS39982-0050EA 133 USD
TS39982-0050 TS39982-0010
3-Nitrosalicylic acid 98%
3-Nitrosalicylic acid
Formula: C₇H₅NO₅
MW: 183.1183 g/mol
Melting Pt: 142…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00024240
CAS Number: 85-38-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR