N-Acetylguanidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Carbamimidoylacetamide1-Acetylguanidine

39744-0050 39744-0250
TS39744-0250EA 279.48 USD
TS39744-0250 TS39744-0050
N-Acetylguanidine 97%
N-Acetylguanidine
Formula: C₃H₇N₃O
MW: 101.11 g/mol
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00075615
CAS Number: 5699-40-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR