(R)-(+)-N,α-Dimethylbenzylamine 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-N-Methyl-1-phenylethanamine
, (R)-N-Methyl-alpha-phenylethylamine
39585-0010
39585-0050
TS39585-0050EA
395
USD
TS39585-0050
TS39585-0010
(R)-(+)-N,α-Dimethylbenzylamine 99+%
(R)-(+)-N,α-Dimethylbenzylamine
Formula:
C₉H₁₃N MW: 135.2062 g/mol Boiling Pt: 74…76 °C (11 mmHg) Flash Pt: 61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044966 CAS Number: 5933-40-4 UN: 2619 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5110 to 1.5130 (20°C, 589 nm) |
GC | ≥99.0 % |
Specific optical rotation | +63° to +67° (20°C, 589 nm) (c=4, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...