Polysorbate 40

Supplier: Thermo Scientific Chemicals

Synonyms: Polyoxyethylene sorbitan monopalmitateSorbitan monopalmitate, ethoxylated

33414-0025 33414-0250 33414-2500 33414-0010
TS33414-0025EA 138 USD
TS33414-0025 TS33414-2500 TS33414-0010 TS33414-0250
Polysorbate 40
Polysorbate 40
Formula: C₆₂H₁₂₂O₂₆
Density: 1.05 g/cm³
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00165345
CAS Number: 9005-66-7

Order Now

Specification Test Results

Appearance Clear to hazy light yellow to dark yellow Viscous liquid or paste
Infrared spectrum Conforms
Acid value ≤3 mg KOH/g
Hydroxyl value 89 to 105 mg KOH/g
Saponification value 41 to 55 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR