8-Pentadecanone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Diheptyl ketoneCaprylone

16979-0250
TS16979-0250EA 716 USD
TS16979-0250
8-Pentadecanone 98%
8-Pentadecanone
Formula: C₁₅H₃₀O
MW: 226.4 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 40…44 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009555
CAS Number: 818-23-5
EINECS: 212-450-7

Order Now

Specification Test Results

Appearance White to light yellow Glistening crystals or flakes
Infrared spectrum Conforms
Melting point 40 to 44 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR