3-Piperidinemethanol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Hydroxymethyl)piperidine

16804-0250 16804-1000 16804-0050
TS16804-0250EA 119.34 USD
TS16804-0250 TS16804-0050 TS16804-1000
3-Piperidinemethanol 98%
3-Piperidinemethanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 236 °C (760 mmHg)
Melting Pt: 58…62 °C
Density: 1.02 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00005997
CAS Number: 4606-65-9
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 56 to 63 °C
GC ≥97.5 %
Iron (Fe) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR