2-Piperidinemethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Hydroxymethyl)piperidine

16803-0050 16803-1000 16803-0250
TS16803-0250EA 170.34 USD
TS16803-0250 TS16803-0050 TS16803-1000
2-Piperidinemethanol 97%
2-Piperidinemethanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 100…102 °C (8hPa)
Melting Pt: 58…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00005987
CAS Number: 3433-37-2
EINECS: 222-333-2

Order Now

Specification Test Results

Appearance White to light orange Solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR