2,6-Diethylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

16522-0010 16522-2500
TS16522-2500EA 57.02 USD
TS16522-2500 TS16522-0010
2,6-Diethylaniline 98%
2,6-Diethylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 3…4 °C
Density: 0.9 g/cm³
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00007753
CAS Number: 579-66-8
EINECS: 209-445-7

Order Now

Specification Test Results

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5446 to 1.5466 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR