2,4,6-Trimethylaniline 97%

Supplier: Thermo Scientific Chemicals
Danger

15796-1000 15796-0025 15796-5000
TS15796-5000EA 157.08 USD
TS15796-5000 TS15796-0025 TS15796-1000
2,4,6-Trimethylaniline 97%
2,4,6-Trimethylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 233 °C (760 mmHg)
Melting Pt: –5 °C
Density: 0.96 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007740
CAS Number: 88-05-1
EINECS: 201-794-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5505 to 1.5525 (20°C, 589 nm)
GC ≥96.0 %
Appearance of solution Clear colorless
Residue after evaporation ≤0.05 %
Related substances ≤0.15 % (Benzaldehyde)
Related substances ≤0.10 % (Cyclohexylmethanol)
Related substances ≤0.04 (Sum of other peaks with relative retention less than that of benzylalcohol)
Related substances ≤0.3 % (Sum of other peaks with relative retention greater than that of benzylalcohol)
Acidity Passes test
Peroxide value ≤5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR