(1S)-(-)-β-Pinene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (-)-Pin-2(10)-ene

15032-0010 15032-2500
TS15032-2500EA 27 USD
TS15032-2500 TS15032-0010
(1S)-(-)-β-Pinene 98%
(1S)-(-)-β-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 165…167 °C (760 mmHg)
Melting Pt: –61 °C
Density: 0.87 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00001345
CAS Number: 18172-67-3
EINECS: 242-060-2
UN: 2319
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4810 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.860 to 0.880
Specific optical rotation -17° to -22° (25°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR