4-Nitrophenyl acetate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Acetoxy-4-nitrobenzene

12875-0250 12875-0050 12875-1000
TS12875-1000EA 161 USD
TS12875-1000 TS12875-0250 TS12875-0050
4-Nitrophenyl acetate 97%
4-Nitrophenyl acetate
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 76…79 °C
Storage Temperature: Freezer
MDL Number: MFCD00007326
CAS Number: 830-03-5
EINECS: 212-593-5

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 76 to 79 °C
GC ≥96.0 %
Specific optical rotation +23° to +29° (20 °C, 589 nm) (c=1 in Chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR