3-Methyl-4-nitrobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methyl-4-nitrobenzoic acid4-Nitro-m-toluic acid

12728-5000 12728-1000
TS12728-1000EA 47.33 USD
TS12728-1000 TS12728-5000
3-Methyl-4-nitrobenzoic acid 99%
4-Nitro-m-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 216…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00007168
CAS Number: 3113-71-1
EINECS: 221-479-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 216 to 219 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR