Dopamine hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,4-Dihydroxyphenethylammonium chloride1-(2-Aminoethyl)-3,4-dihydroxybenzene HCl3-Hydroxytyramine hydrochloride3-Hydroxytyraminium chloride

12200-0100 12200-1000 12200-0250
TS12200-0250EA 110.65 USD
TS12200-0250 TS12200-1000 TS12200-0100
Dopamine hydrochloride 99%
Dopamine hydrochloride
Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Melting Pt: 241…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00012898
CAS Number: 62-31-7
EINECS: 200-527-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 2 hrs)
Sulfated ash ≤0.1 %
Titration with HClO4 ≥98.5 % (On dried substance)
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR