3-Chloro-1H-pyrrolo[2,3-b]pyridine

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 3-Chloro-7-azaindole

OR14108-1G
76615-682EA 20 USD
76615-682
3-Chloro-1H-pyrrolo[2,3-b]pyridine
3-Chloro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD08272228
CAS Number: 80235-01-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR