4-(4-Methylpiperazino)-1,2-benzenediamine 95%

Supplier: APOLLO SCIENTIFIC

OR322770-1G OR322770-250MG
76616-460EA 74 USD
76616-460 76616-458
4-(4-Methylpiperazino)-1,2-benzenediamine 95%
4-(4-Methylpiperazino)-1,2-benzenediamine
Formula: C₁₁H₁₈N₄
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD03001209
CAS Number: 54998-08-2
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR