6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
6-Chloro-[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one
OR111545-5G
OR111545-10G
OR111545-1G
76624-716EA
148
USD
76624-716
76624-712
76624-714
6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one
6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one
Formula:
C₅H₃ClN₄O MW: 170.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01219639 CAS Number: 33050-32-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...