2-Amino-3-cyano-4,5-di(furyl)furan

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 5'-Amino-2,2':3',2''-terfuran-4'-carbonitrile

OR01757-5G OR01757-1G OR01757-250MG
76624-640EA 42 USD
76624-640 76624-642 76624-638
2-Amino-3-cyano-4,5-di(furyl)furan
2-Amino-3-cyano-4,5-di(furyl)furan
Formula: C₁₃H₈N₂O₃
MW: 240.22 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00099027
CAS Number: 24386-17-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR