1-(Bromomethyl)adamantane 95+%

Supplier: APOLLO SCIENTIFIC

Danger

OR018152-10G OR018152-1G OR018152-5G
76615-190EA 260 USD
76615-190 76615-192 76615-194
1-(Bromomethyl)adamantane 95+%
1-(Bromomethyl)adamantane
Formula: C₁₁H₁₇Br
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD00185303
CAS Number: 14651-42-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR