Methyl-4-Chloro-3-methylbenzoate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 4-chloro-3-methylbenzoate4-Chloro-3-methylbenzoic Acid Methyl Ester

OR5430-1G
76591-778EA 13.39 USD
76591-778
Methyl-4-Chloro-3-methylbenzoate
Methyl-4-Chloro-3-methylbenzoate
Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 78 °C/0.06mmHg
Density: 1.21 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151838
CAS Number: 91367-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR