Tetrahydro-2H-pyran-4-carbonyl chloride 97%

Supplier: APOLLO SCIENTIFIC

Danger

OR14254-5G OR14254-1G OR14254-10G
76620-684EA 74 USD
76620-684 76620-682 76620-680
Tetrahydro-2H-pyran-4-carbonyl chloride 97%
Tetrahydro-2H-pyran-4-carbonyl chloride
Formula: C₆H₉ClO₂
MW: 148.5875 g/mol
Boiling Pt: 50…52 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD06200863
CAS Number: 40191-32-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR