3-(Chlorosulfonyl)benzoic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR21680-5G
76616-018EA 13.39 USD
76616-018
3-(Chlorosulfonyl)benzoic acid
3-(Chlorosulphonyl)benzoic acid
Formula: C₇H₅ClO₄S
MW: 220.63 g/mol
Melting Pt: 128…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00024877
CAS Number: 4025-64-3
EINECS: 223-692-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR