2-(1-Pyrrolyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR1437-1G OR1437-5G
76624-798EA 41 USD
76624-798 76624-800
2-(1-Pyrrolyl)benzonitrile
2-(1-Pyrrolyl)benzonitrile
Formula: C₁₁H₈N₂
Storage Temperature: Ambient
MDL Number: MFCD00128308
CAS Number: 33265-71-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR