4,6-Dichloro-2-(methylthio)pyrimidine

Supplier: APOLLO SCIENTIFIC

Danger

OR29949-25G OR29949-100G OR29949-500G
76620-996EA 81 USD
76620-996 76621-000 76620-998
4,6-Dichloro-2-(methylthio)pyrimidine
4,6-Dichloro-2-(methylthio)pyrimidine
Formula: C₅H₄Cl₂N₂S
MW: 195.07 g/mol
Boiling Pt: 135…136 °C/14mmHg
Melting Pt: 38…42 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006086
CAS Number: 6299-25-8
EINECS: 228-577-6
UN: 3263
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR