4-(3-Cyanophenyl)morpholine 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Morpholinobenzonitrile3-(Morpholin-4-yl)benzonitrile

OR4431-1G
76625-200EA 22 USD
76625-200
4-(3-Cyanophenyl)morpholine 97%
4-(3-Cyanophenyl)morpholine
Formula: C₁₁H₁₂N₂O
MW: 188.2300 g/mol
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD07368523
CAS Number: 204078-31-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR