5-(Tributylstannyl)thiazole

Supplier: APOLLO SCIENTIFIC

Danger

OR15547-5G OR15547-1G OR15547-250MG
76611-164EA 31 USD
76611-164 76611-166 76611-162
5-(Tributylstannyl)thiazole
5-(Tributylstannyl)thiazole
Formula: C₁₅H₂₉NSSn
Storage Temperature: Refrigerator
MDL Number: MFCD01319074
CAS Number: 157025-33-7
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR