1-Methyl-5-(tributylstannyl)imidazole, Technical Grade

Supplier: APOLLO SCIENTIFIC

Danger

OR4409-250MG OR4409-5G OR4409-1G
76611-860EA 196 USD
76611-860 76611-856 76611-858
1-Methyl-5-(tributylstannyl)imidazole, Technical Grade
1-Methyl-5-(tributylstannyl)imidazole
Formula: C₁₆H₃₂N₂Sn
MW: 371.15
Boiling Pt: 160…164 °C/2mmHg
Density: 1.22 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01319031
CAS Number: 147716-03-8
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR