6-Bromo-1-chloroisoquinoline

Supplier: APOLLO SCIENTIFIC

Danger

OR300053-5G OR300053-1G
76616-302EA 179 USD
76616-302 76616-300
6-Bromo-1-chloroisoquinoline
6-Bromo-1-chloroisoquinoline
Formula: C₉H₅BrClN
MW: 242.5000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738661
CAS Number: 205055-63-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR