4,6-Dichloroquinoline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5287-5G OR5287-1G
76591-386EA 50 USD
76591-386 76591-388
4,6-Dichloroquinoline 98%
4,6-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00156141
CAS Number: 4203-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR