5-Chloro-2-hydroxybenzonitrile 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR24196-5G OR24196-1G
76624-970EA 24.72 USD
76624-970 76624-972
5-Chloro-2-hydroxybenzonitrile 95+%
5-Chloro-2-hydroxybenzonitrile
Formula: C₇H₄ClNO
MW: 153.5700 g/mol
Melting Pt: 152…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00219003
CAS Number: 13589-72-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR