7-Chloro-1H-indazole

Supplier: APOLLO SCIENTIFIC

Danger

OR3680-1G OR3680-5G OR3680-250MG
76616-526EA 244 USD
76616-526 76616-524 76616-522
7-Chloro-1H-indazole
7-Chloro-1H-indazole
Formula: C₇H₅ClN₂
MW: 152.5810 g/mol
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD07368214
CAS Number: 37435-12-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR