3,5-Dimethyl-4-hydroxybenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Hydroxy-3,5-dimethylbenzonitrile

OR26857-5G OR26857-25G OR26857-100G
76616-196EA 13.39 USD
76616-196 76616-194 76616-192
3,5-Dimethyl-4-hydroxybenzonitrile
3,5-Dimethyl-4-hydroxybenzonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00221716
CAS Number: 4198-90-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR