3-Amino-4-bromo-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-1H-indazol-3-amine

OR30982-5G OR30982-1G
76616-408EA 32 USD
76616-408 76616-410
3-Amino-4-bromo-1H-indazole
3-Amino-4-bromo-1H-indazole
Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD09909580
CAS Number: 914311-50-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR