5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC

Danger

OR23902-5G OR23902-1G OR23902-250MG
76620-832EA 74 USD
76620-832 76620-830 76620-828
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonyl chloride
Formula: C₅H₆Cl₂N₂O₂S
Boiling Pt: 105 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052536
CAS Number: 88398-93-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR