Methyl vanillate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 4-hydroxy-3-methoxybenzoate

OR5261-25G OR5261-500G OR5261-100G
76591-292EA 17 USD
76591-292 76591-294 76591-296
Methyl vanillate
Methyl vanillate
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 285…287 °C
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008438
CAS Number: 3943-74-6
EINECS: 223-525-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR