(4-tert-Butylphenyl)methanesulfonyl chloride 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
4-tert-Butylbenzylsulphonyl chloride
OR300728-1G
OR300728-5G
76621-006EA
82
USD
76621-006
76621-008
(4-tert-Butylphenyl)methanesulfonyl chloride 95%
(4-tert-Butylphenyl)methanesulphonyl chloride
Formula:
C₁₁H₁₅ClO₂S Storage Temperature: Ambient |
UN:
3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...