(3,5-Dichlorophenyl)methanesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3,5-Dichloro-α-toluenesulphonyl chloride

OR12682-1G OR12682-250MG
76615-576EA 70 USD
76615-576 76615-578
(3,5-Dichlorophenyl)methanesulfonyl chloride
(3,5-Dichlorophenyl)methanesulphonyl chloride
Formula: C₇H₅CI₃O₂S
MW: 259.54 g/mol
Melting Pt: 108…111 °C
Storage Temperature: Ambient
UN: 3096
ADR: 8+4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR