(R)-(-)-Glycidyl butyrate

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: (-)-[(2R)-Oxiran-2-yl]methyl butanoate(R)-Oxiran-2-ylmethyl butyrate

OR28822-25G OR28822-100G OR28822-5G
76611-494EA 13 USD
76611-494 76611-490 76611-492
(R)-(-)-Glycidyl butyrate
(R)-(-)-Glycidyl butyrate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 197 °C
Density: 1.032 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00075120
CAS Number: 60456-26-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR