ω-Bromo-2,5-dichloroacetophenone

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 2-Bromo-1-(2,5-dichlorophenyl)ethanone

OR47840-5G OR47840-1G OR47840-25G
76611-928EA 175 USD
76611-928 76611-924 76611-926
ω-Bromo-2,5-dichloroacetophenone
ω-Bromo-2,5-dichloroacetophenone
Formula: C₈H₅BrCl₂O
Melting Pt: 33…35 °C
Storage Temperature: Ambient
CAS Number: 4571-25-9
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR