2-Bromo-4'-(1-pyrrolidinyl)acetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Pyrrolidin-1-yl)phenacyl bromide

OR11500-5G OR11500-1G OR11500-25G
76615-476EA 410 USD
76615-476 76615-478 76615-474
2-Bromo-4'-(1-pyrrolidinyl)acetophenone 97%
2-Bromo-4'-(1-pyrrolidinyl)acetophenone
Formula: C₁₂H₁₄BrNO
MW: 268.16
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD01075690
CAS Number: 216144-18-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR