5-Bromo-2-chloroaniline

Supplier: APOLLO SCIENTIFIC

Warning

OR0859-5G OR0859-25G
76615-270EA 41 USD
76615-270 76615-272
5-Bromo-2-chloroaniline
5-Bromo-2-chloroaniline
Formula: C₆H₅BrClN
MW: 206.47 g/mol
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD06796283
CAS Number: 60811-17-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR