2-Chloronicotinoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR21417-5G OR21417-100G OR21417-25G
76616-006EA 18 USD
76616-006 76616-004 76616-002
2-Chloronicotinoyl chloride 97%
2-Chloronicotinoyl chloride
Formula: C₆H₃Cl₂NO
MW: 176.00 g/mol
Boiling Pt: 98…100 °C/2mmHg
Melting Pt: 39…45 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051677
CAS Number: 49609-84-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR