(R)-1-Boc-3-Hydroxypiperidine 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate(R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate(R)-1-BOC-3-hydroxypiperidine

OR4584-25G OR4584-1G OR4584-5G
76616-800EA 16 USD
76616-800 76616-798 76616-796
(R)-1-Boc-3-Hydroxypiperidine 98%
(R)-1-Boc-3-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 43…50 °C
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD04115306
CAS Number: 143900-43-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR