Bis(pinacolato)diboron 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane)
, 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
OR5456-500G
OR5456-25G
OR5456-100G
76591-990EA
24.72
USD
76591-990
76591-992
76591-994
Bis(pinacolato)diboron 99%
Bis(pinacolato)diboron
Versatile reagent for the preparation of boronic esters:J.Org.Chem.1995, 60,7508-7510.
Formula:
C₁₂H₂₄B₂O₄ MW: 253.94 g/mol Melting Pt: 137…140 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00799570 CAS Number: 73183-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...