1,3,5-Tris(bromomethyl)benzene 97+%

Supplier: APOLLO SCIENTIFIC

Danger

OR18429-5G OR18429-1G OR18429-25G
76615-930EA 33 USD
76615-930 76615-932 76615-934
1,3,5-Tris(bromomethyl)benzene 97+%
1,3,5-Tris(bromomethyl)benzene
Formula: C₉H₉Br₃
Melting Pt: 94…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00182500
CAS Number: 18226-42-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR