3-Bromo-2-thiophenesulfonyl chloride 90%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 3-Bromothiophene-2-sulfonyl chloride

OR103634-1G OR103634-50MG OR103634-250MG
76615-316EA 133 USD
76615-316 76615-318 76615-320
3-Bromo-2-thiophenesulfonyl chloride 90%
3-Bromo-2-thiophenesulphonyl chloride
Formula: C₄H₂BrClO₂S₂
Melting Pt: 39…41.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08059484
CAS Number: 170727-02-3
UN: 2923
ADR: 8+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR