2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Isopropyl pinacolyl borate4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane

OR10991-500G OR10991-100G OR10991-25G
76610-992EA 133 USD
76610-992 76610-990 76610-988
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₉H₁₉BO₃
MW: 186.06 g/mol
Boiling Pt: 73 °C/15mmHg
Density: 0.912 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00192241
CAS Number: 61676-62-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR