1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Benzyl-4-hydroxypiperidine-4-carbonitrile hydrochloride

OR5395-5G OR5395-25G OR5395-100G
76625-302EA 13 USD
76625-302 76625-300 76625-298
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride 98%
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride
Formula: C₁₃H₁₇ClN₂O
Melting Pt: 176 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012836
CAS Number: 71617-20-4
EINECS: 275-713-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR