Ethyl cyclopropanecarboxylate 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR5300-100G OR5300-25G
76625-288EA 13 USD
76625-288 76625-286
Ethyl cyclopropanecarboxylate 98%
Ethyl cyclopropanecarboxylate
Formula: C₆H₁₀O₂
MW: 114.15 g/mol
Boiling Pt: 133…134 °C
Density: 0.96 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
EINECS: 225-010-4
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR