Sulfur trioxide trimethylamine complex 95%

Supplier: APOLLO SCIENTIFIC

Danger

OR5415-500G OR5415-25G OR5415-100G
76621-638EA 342 USD
76621-638 76621-634 76621-636
Sulfur trioxide trimethylamine complex 95%
Sulphur trioxide trimethylamine complex
Formula: C₃H₉NO₃S
Melting Pt: 232 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012421
EINECS: 221-614-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR