1-(3-Acetylphenyl)-2-thiourea

Supplier: APOLLO SCIENTIFIC

Warning

OR1414-10G OR1414-50G
76624-782EA 226 USD
76624-782 76624-784
1-(3-Acetylphenyl)-2-thiourea
1-(3-Acetylphenyl)-2-thiourea
Formula: C₉H₁₀N₂OS
Melting Pt: 165…167 °C
Storage Temperature: Ambient
UN: 3335
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR